Kimble™ KIMAX™ Burets with Threaded Closure Fit
Kimble™ KIMAX™ Burets with Threaded Closure Fit
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Burets with Threaded Closure Fit
Educational grade
Have Questions?
Catalog NumberCapacity (Metric)
FIS03-857B50 mL
Catalog number FIS03-857B
Price (SGD)
413.00
CS
Add to cart
Capacity (Metric):
50 mL
Price (SGD)
413.00
CS
Add to cart
  • Economical and versatile
  • Accept PTFE stopcock assembly with threaded closure to prevent dislodging
  • Tube has a reinforced top with tooled lower end
  • Ground tip fits into lower end of stopcock assembly
  • Quantity: 1/Cs.
Specifications
Capacity (Metric)50 mL
Length (Metric)725 mm
MaterialBorosilicate Glass
Graduations0.1 mL
For Use With (Application)Educational Institutions and for Many General Laboratory Procedures
Length (English)28.54 in.
IncludesDust Cap
ScaleBlack
Stopcock MaterialPTFE
TypeEducational Grade Buret
ClassClass A
Certifications/ComplianceASTM E438
Unit SizeCS