Kimble™ KIMAX™ Burets with PTFE-Plug Stopcock
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Burets with PTFE-Plug Stopcock
Class B
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS03-700-20B25 mL
FIS03-700-20C50 mL
Catalog number FIS03-700-20B
Price (SGD)
488.00
Each
Capacity (Metric):
25 mL
Price (SGD)
488.00
Each
Class B
  • Permanently colored black scale
If glass body breaks, stopcock plug can be used in replacement glass body (03-700-21C, sold separately).
Specifications
Capacity (Metric)25 mL
Length (Metric)571 mm
MaterialBorosilicate Glass
Graduations0.1 mL
For Use With (Application)General Purpose Volumetric Analysis and Titrations where Class B Tolerances are Appropriate
Length (English)22.48 in.
IncludesDust Cap
ScaleBlack
StopcockStraight
Stopcock MaterialPTFE
Tolerance±0.06 mL
TypeBore Buret
ClassClass B
Certifications/ComplianceASTM E287, ASTM E438, ASTM E542, ASTM E694
Unit SizeEach