Brass Wire Brush
Brass Wire Brush
Actual product may vary
Brass Wire Brush
For cleaning corroded or dirty metal surfaces
Have Questions?
Catalog NumberLength (English)
FIS03-68510 in.
Catalog number FIS03-685
Price (SGD)
83.00
EA
Add to cart
Length (English):
10 in.
Price (SGD)
83.00
EA
Add to cart
  • Wooden handle
  • Handle length: 5 in. (13cm)
Specifications
Product TypeBrush
Handle MaterialWood
For Use With (Application)For cleaning corroded or dirty metal surfaces
Length (English)10 in.
Length (English) Bristle5 in.
Length (English) Handle5 in.
Length (Metric)25.4 cm
Length (Metric) Bristle12.7 cm
Length (Metric) Handle13 cm
Width (English) Bristle0.62 in.
Width (Metric) Bristle1.6 cm
Unit SizeEA