Table-Dusting Brushes
Table-Dusting Brushes
Actual product may vary
Table-Dusting Brushes
For dusting tables and lab benches
Have Questions?
Catalog NumberLength (English)
FIS03-68012.62 in.
Catalog number FIS03-680
Price (SGD)
71.00
EA
Add to cart
Length (English):
12.62 in.
Price (SGD)
71.00
EA
Add to cart
  • Soft-bristle brush
  • Handle, 5 in.L (13cm)
Specifications
Product TypeSoft Bristle Brush
For Use With (Application)For dusting tables and lab benches
Length (English)12.62 in.
Length (English) Bristle7.5 in.
Length (English) Handle5 in.
Length (Metric)32 cm
Length (Metric) Bristle19 cm
Length (Metric) Handle13 cm
Width (English) Bristle2 in.
Width (Metric) Bristle5 cm
Unit SizeEA