Flat Camel's Hair Brushes
Flat Camel's Hair Brushes
Actual product may vary
Flat Camel's Hair Brushes
Flat Camel's Hair Brushes are useful for dusting sensitive instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric) BristleWidth (Metric) Bristle
FIS03-665D3.3 cm5.0 cm
FIS03-665A1.9 cm1.3 cm
FIS03-665B2.5 cm2.5 cm
Catalog number FIS03-665D
Price (SGD)
60.00
EA
Add to cart
Length (Metric) Bristle:
3.3 cm
Width (Metric) Bristle:
5.0 cm
Price (SGD)
60.00
EA
Add to cart
Flat Camel's Hair Brushes are useful for dusting sensitive instruments.
  • Hair mounted with metal ferrule
  • Wooden handle
Specifications
Product TypeFlat Brush
Bristle MaterialCamel Hair
Handle MaterialWood
For Use With (Application)For dusting sensitive instruments
IncludesMetal ferrule
Length (English)7 in.
Length (English) Bristle1.31 in.
Length (Metric)18 cm
Length (Metric) Bristle3.3 cm
Width (English) Bristle2 in.
Width (Metric) Bristle5.0 cm
Unit SizeEA