Check Order Status
Long-Handled Camel's Hair Brush
Actual product may vary
Long-Handled Camel's Hair Brush
For dusting balances.
Have Questions?
Catalog Number
Color
FIS03-661
Black
Catalog number
FIS03-661
Price (SGD)
34.00
EA
Add to cart
Color:
Black
Price (SGD)
34.00
EA
Add to cart
For dusting balances.
Slightly pointed tip and full body tapering toward handle.
Specifications
Product Type
Brush
Bristle Material
Camel Hair
Tip Type
Pointed
Color
Black
Diameter (English) Bristle
0.75 in.
Diameter (Metric) Bristle
1.9 cm
For Use With (Application)
For dusting balances
Length (English)
9 in.
Length (English) Bristle
1.5 in.
Length (Metric)
23 cm
Length (Metric) Bristle
3.8 cm
Size
Small