Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Facility Safety and Maintenance
▸
Hand Tools and Power Tools
▸
Brushes
▸
Pipe Cleaner Brushes
Actual product may vary
BJ Long
Pipe Cleaner Brushes
Cut at any length for cleaning small pipets, stopcocks, capillary tubes
Have Questions?
Change view
Catalog Number
For Use With (Equipment)
FIS03-642B
0.098 to 0.196 in. O.D. Tubing
FIS03-642A
0.059 to 0.11 in. O.D. Tubing
2 Options
Catalog number
FIS03-642B
Price (SGD)
-
For Use With (Equipment):
0.098 to 0.196 in. O.D. Tubing
Cut at any length for cleaning small pipets, stopcocks, capillary tubes
May be used for slightly larger sizes by bending in moderate zig-zag pattern
Specifications
Product Type
Pipe Cleaner Brush
Handle Material
Galvanized Wire
For Use With (Application)
For cleaning small pipets, stopcocks, capillary tubes
For Use With (Equipment)
0.098 to 0.196 in. O.D. Tubing
Length (English)
53 ft.
Length (Metric)
16.2 m