Pipe Cleaner Brushes
Actual product may vary
BJ Long
Pipe Cleaner Brushes
Cut at any length for cleaning small pipets, stopcocks, capillary tubes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
FIS03-642A0.059 to 0.11 in. O.D. Tubing
FIS03-642B0.098 to 0.196 in. O.D. Tubing
Catalog number FIS03-642A
Price (SGD)
-
For Use With (Equipment):
0.059 to 0.11 in. O.D. Tubing
Cut at any length for cleaning small pipets, stopcocks, capillary tubes
  • May be used for slightly larger sizes by bending in moderate zig-zag pattern
Specifications
Product TypePipe Cleaner Brush
For Use With (Application)For cleaning small pipets, stopcocks, capillary tubes
For Use With (Equipment)0.059 to 0.11 in. O.D. Tubing
Length (English)53 ft.
Length (Metric)16.2 m
Unit SizePack