Soft Nylon Bristle Brush
Soft Nylon Bristle Brush
Actual product may vary
Fisherbrand
Soft Nylon Bristle Brush
For cleaning funnel stems and capillary tubes.
Have Questions?
Catalog NumberColor
FIS03-636White
Catalog number FIS03-636
Price (SGD)
53.00
PK
Add to cart
Color:
White
Price (SGD)
53.00
PK
Add to cart
For cleaning funnel stems and capillary tubes.
  • Made from soft nylon bristles Length: 12 in. (30cm)
Specifications
Product TypeSoft Bristle Brush
Bristle MaterialNylon
Bristle TypeSoft
ColorWhite
Diameter (English) Bristle0.5 in.
Diameter (Metric) Bristle1.3 cm
For Use With (Application)For cleaning funnel stems and capillary tubes
Length (English)12 in.
Length (English) Bristle2.5 in.
Length (Metric)30.5 cm
Length (Metric) Bristle6.3 cm
SizeMedium
Unit SizePK