Stiff Bristled Brushes
Stiff Bristled Brushes
Actual product may vary
Stiff Bristled Brushes
For cleaning tubes of 0.43 in. dia. (1.1cm).
Have Questions?
Catalog NumberColor
FIS03-635Natural
Catalog number FIS03-635
Price (SGD)
51.00
PK
Add to cart
Color:
Natural
Price (SGD)
51.00
PK
Add to cart
  • Overall Length: 12 in. (30cm)
  • Bristles: 3L x 0.5 in. dia. (7.6 x 1.3cm)
Specifications
Product TypeBrush
ColorNatural
Diameter (English) Bristle0.5 in.
Diameter (Metric) Bristle1.3 cm
For Use With (Application)For cleaning 0.43 in. dia. tubes
Length (English)12 in.
Length (English) Bristle3 in.
Length (Metric)30.5 cm
Length (Metric) Bristle7.6 cm
SizeMedium
Unit SizePK