Nessler Tube Brushes, Natural black bristle
Nessler Tube Brushes, Natural black bristle
Actual product may vary
Nessler Tube Brushes, Natural black bristle
Fisherbrand™ Nessler Tube Brushes are for use with low- and high-form Nessler tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Length (Metric) BristleFor Use With (Equipment)
FIS03-62248 cm10 cmFor high-form nessler tubes
FIS03-621C76 cm15.2 cmFor low-form nessler tubes
FIS03-621B50.8 cm15.2 cmFor low-form nessler tubes
FIS03-621A27.94 cm10 cmFor low-form nessler tubes
Catalog number FIS03-622
Price (SGD)
-
Length (Metric):
48 cm
Length (Metric) Bristle:
10 cm
For Use With (Equipment):
For high-form nessler tubes
Fisherbrand™ Nessler Tube Brushes are for use with low- and high-form Nessler tubes.
  • Full bristles to make complete contact with cylinder walls and bottom
  • 03-621A is natural black bristle; others are black nylon
  • Radial, double-fan tip
  • Galvanized wire handle
Specifications
Product TypeNessler Tube Brush
Bristle MaterialNylon
Handle MaterialGalvanized Wire
Tip TypeRadial, Double Fan Tip
ColorBlack
Diameter (English) Bristle1 in.
Diameter (Metric) Bristle2.54 cm
For Use With (Equipment)For high-form nessler tubes
Length (English)19 in.
Length (English) Bristle4 in.
Length (Metric)48 cm
Length (Metric) Bristle10 cm
SizeLarge
Unit SizeEA