Check Order Status
Buret Brushes
Actual product may vary
Fisherbrand
Buret Brushes
Fisherbrand™ Buret Brushes feature a wire handle galvanized to resist rust and corrosion.
Have Questions?
Catalog Number
For Use With (Equipment)
Length (Metric) Bristle
Diameter (Metric) Bristle
FIS03-615
75 to 100mL buret
10.2 cm
2.5 cm
Catalog number
FIS03-615
Price (SGD)
19.00
EA
Add to cart
For Use With (Equipment):
75 to 100mL buret
Length (Metric) Bristle:
10.2 cm
Diameter (Metric) Bristle:
2.5 cm
Price (SGD)
19.00
EA
Add to cart
Fisherbrand™ Buret Brushes feature a wire handle galvanized to resist rust and corrosion.
03-614 features tapered bristle section
Specifications
Product Type
Buret Brush
Bristle Type
Tapered
Handle Material
Galvanized Wire
Color
Black
Diameter (English) Bristle
1 in.
Diameter (Metric) Bristle
2.5 cm
For Use With (Equipment)
75 to 100mL buret
Length (English)
36 in.
Length (English) Bristle
4 in.
Length (Metric)
91.5 cm
Length (Metric) Bristle
10.2 cm
Size
Large