Check Order Status
Flexible-Handle Brushes
Actual product may vary
Fisherbrand
Flexible-Handle Brushes
Soft, single row of bristles fits easily into flask necks
Have Questions?
Change view
Catalog Number
Length (Metric)
For Use With (Equipment)
FIS03-570C
43.18 cm
1000mL flasks
FIS03-570A
30.5 cm
250mL flasks
2 Options
Catalog number
FIS03-570C
Price (SGD)
59.00
EA
Add to cart
Length (Metric):
43.18 cm
For Use With (Equipment):
1000mL flasks
Price (SGD)
59.00
EA
Add to cart
Designed for cleaning Erlenmeyer, filtering, Kjeldahl, Nephelo, pear-shaped, round-bottom, flat-bottom, reaction and volumetric flasks.
Plastic handle bends to allow fitting of bristles to container shape
Handle has hanging ring
Specifications
Product Type
Flask Brush
Bristle Type
Soft
Handle Material
Plastic
Handle Type
Flexible
Diameter (English) Bristle
1.75 in.
Diameter (Metric) Bristle
4.4 cm
For Use With (Equipment)
1000mL flasks
Includes
Hanging ring
Length (English)
17 in.
Length (English) Bristle
5 in.
Length (Metric)
43.18 cm
Length (Metric) Bristle
12.7 cm
No. of Bristle Rows
1