Beakers Brush
Beakers Brush
Actual product may vary
Fisherbrand
Beakers Brush
Brush for cleaning beakers
Have Questions?
Catalog NumberColor
FIS03-555Black
Catalog number FIS03-555
Price (SGD)
83.00
EA
Add to cart
Color:
Black
Price (SGD)
83.00
EA
Add to cart
Brush with a wood handle for cleaning beakers
  • Two rows of bristles provide slightly oval shape
  • End tuft for cleaning corners
  • Wooden handle
Specifications
Product TypeBrush
Bristle TypeTufted End
Handle MaterialWood
Tip TypeTuft End
ColorBlack
Diameter (English) Bristle3 in.
Diameter (Metric) Bristle7.6 cm
For Use With (Application)For beakers
Length (English)16 in.
Length (English) Bristle6 in.
Length (Metric)40.64 cm
Length (Metric) Bristle15.24 cm
No. of Bristle Rows2
SizeLarge
Unit SizeEA