Brush with Four Bristle Rows
Brush with Four Bristle Rows
Actual product may vary
Brush with Four Bristle Rows
With a double-tufted end for cleaning beakers.
Have Questions?
Catalog NumberLength (English)
FIS03-54513 in.
Catalog number FIS03-545
Price (SGD)
61.00
EA
Add to cart
Length (English):
13 in.
Price (SGD)
61.00
EA
Add to cart
With a double-tufted end for cleaning beakers.
  • Wooden handle
  • Length: 13 in.
Specifications
Product TypeBeaker Brush
Bristle TypeDoubleTufted
Handle MaterialWood
ColorBlack
Diameter (English) Bristle3.25 in.
Diameter (Metric) Bristle8 cm
For Use With (Application)For cleaning beakers
Length (English)13 in.
Length (English) Bristle5.5 in.
Length (Metric)33.02 cm
Length (Metric) Bristle14 cm
No. of Bristle Rows4
SizeSmall
Unit SizeEA