White Nylon Bristle Brush
White Nylon Bristle Brush
Actual product may vary
Fisherbrand
White Nylon Bristle Brush
Fisherbrand™ White Nylon Bristle Brushes are heavy-duty brushes for cleaning beakers, jars, and similar containers.
Have Questions?
Catalog NumberColor
FIS03-536White
Catalog number FIS03-536
Price (SGD)
212.00
EA
Add to cart
Color:
White
Price (SGD)
212.00
EA
Add to cart
Fisherbrand™ White Nylon Bristle Brushes are heavy-duty brushes for cleaning beakers, jars, and similar containers.
  • Extra-heavy bristle with double-tufted end
  • Wooden handle
  • Chemical-resistant
Specifications
Product TypeBristle Brush
Bristle MaterialNylon
Bristle TypeDoubleTufted
Handle MaterialWood
Tip TypeDouble Tufted End
ColorWhite
Diameter (English) Bristle2.87 in.
Diameter (Metric) Bristle7.2 cm
For Use With (Application)For cleaning beakers, jars, and similar containers
Length (English)16 in.
Length (English) Bristle5 in.
Length (Metric)40.64 cm
Length (Metric) Bristle12.7 cm
SizeMedium
Unit SizeEA