Spacers for SE 600 Chroma, SE 600 Standard, or SE 400 Vertical Electrophoresis Units
Actual product may vary
Hoefer
Spacers for SE 600 Chroma, SE 600 Standard, or SE 400 Vertical Electrophoresis Units
Accessory parts for SE 600 Chroma SE 600 Standard or SE 400 Vertical Electrophoresis Units
Have Questions?
Catalog NumberThickness (Metric)Width (Metric)Gel Dimensions
FIS03-500-2100.1 cm2 cm2 x 16 cm x 1.0mm
Catalog number FIS03-500-210
Price (SGD)
-
Thickness (Metric):
0.1 cm
Width (Metric):
2 cm
Gel Dimensions:
2 x 16 cm x 1.0mm
  • Ranges from 0.75 to 3.0 mm Thickness
  • 8, 16 and 24 cm in Length
  • 1 to 2 cm Width
  • 2 Pk.
Specifications
For Use With (Equipment)SE 600 Chroma, SE 600 Standard, SE 400 Vertical Electrophoresis Unit
TypeSpacer
Gel Dimensions2 x 16 cm x 1.0mm
Quantity2/Pk.
Thickness (English)0.03 in.
Thickness (Metric)0.1 cm
Length (Metric)16 cm
Width (Metric)2 cm
Unit SizeEach