True North™ Polypropylene Film Freezer Box, 1.5/2 mL, 81-Place
Actual product may vary
Fisherbrand
True North™ Polypropylene Film Freezer Box, 1.5/2 mL, 81-Place
The True North™ Freezer Box is reusable, economical and constructed of moisture resistant thin polypropylene. They are supplied 'flat-packed' eliminating the hassle of stacking and reducing the amount of space needed for storage, and shipping costs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FIS03-489-126Light Green
FIS03-489-124White
FIS03-489-125Blue
FIS03-489-127Purple
Catalog number FIS03-489-126
Price (SGD)
274.00
Pack
Color:
Light Green
Price (SGD)
274.00
Pack
  • Easy self-assemble boxes are shipped flat
  • More durable than cardboard boxes as they are moisture resistant
  • Dividers included with the boxes
  • Corrugated Polypropylene: Store down to temperatures of -80°C.
  • Must use tubes capable of being stored at the required temperature of the user
  • Not recommended for autoclaving
  • Mold Resistant
Specifications
Capacity (Metric)1.5 to 2.0 mL
DescriptionTrue North™ Polypropylene Film Freezer Box, 1.5/2 mL, 81-Place, Lt. Green
ColorLight Green
MaterialPolypropylene
ShapeRectangular
TypeFreezer Box
For Use With (Application)Tube Storage
For Use With (Equipment)Tubes, Freezer Racks, Freezers
Holds81 (1.5 and 2 mL Tubes)
IncludesDividers
Max. Temperature (English)-112°F
Max. Temperature (Metric)-80°C
Min. Temperature (English)-320.8°F
Min. Temperature (Metric)-196°C
Product LineTrue North
Quantity10 Pk.
Withstand Temperature (Metric)-196°C
Dimensions (L x W)5 x 4.9 in.
Dimensions (L x W x H)5 x 4.9 x 1.9 in.
Length (English)4.95 in.
Length (Metric)12.5 cm
Width (English)4.92 in.
Width (Metric)12.4 cm
Height (English)1.92 in.
Height (Metric)4.9 cm
Diameter (English) Well0.5 in.
Diameter (Metric) Well12.1 mm
Weight (English)0.2 lb.
Weight (Metric)0.09 kg
Unit SizePack