Nalgene™ Utility Boxes
Nalgene™ Utility Boxes
Actual product may vary
Nalgene™ Utility Boxes
Nalgene™ Utility Boxes
Nalgene™ Utility Boxes
Thermo Scientific
Nalgene™ Utility Boxes
Use these attractive, easy-to-clean boxes, available in four different sizes and shapes, for general-purpose laboratory storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS03-484D1000 mL
FIS03-484B325 mL
FIS03-484C500 mL
Catalog number FIS03-484D
Price (SGD)
-
Capacity (Metric):
1000 mL
Use these attractive, easy-to-clean Thermo Scientific™ Nalgene™ Utility Boxes for general-purpose laboratory storage. Ideal for multi-purpose lab storage, boxes are available in four different sizes and shapes. Lids snap securely in place, clear walls offer easy view of contents and rounded corners allow for easy cleaning.
  • Manufactured from clear styrene-acrylonitrile with tight-fitting, molded, white, LDPE lids that include inserts to store and keep items dry and secure
  • Attractive and easy to clean
  • Stackable for high-density storage
  • Cat. No. 5700-0500 is ideal for electrophoresis gels
  • Not leakproof
Specifications
Capacity (English)32 oz.
Capacity (Metric)1000 mL
Closure ColorWhite
Closure MaterialLDPE
DescriptionSample Box SAN 1000mL
Closure TypeLid
ColorClear
MaterialSAN
ShapeRectangular
For Use With (Application)Storage
IncludesInsert
Mounting TypeStackable
Product TypeStorage Box
Quantity2/Pk.; 12/Cs.
Dimensions (L x W x H)5 x 2.9 x 6.9 in. (13 x 7 x 18 cm)
Length (Metric)13 cm
Length (English)5 in.
Width (Metric)7 cm
Width (English)2.9 in.
Height (Metric)18 cm
Height (English)6.9 in.
Unit SizePK