Check Order Status
Kimble™ KIMAX™ Weighing Bottles — With standard taper Outside Ground Caps
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Weighing Bottles — With standard taper Outside Ground Caps
Tall, cylindrical form with standard taper caps
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Standard Taper Joint
FIS03-422H
92 mL
45/12
FIS03-422E
30 mL
34/12
2 Options
Catalog number
FIS03-422H
Price (SGD)
2,067.00
PK
Add to cart
Capacity (Metric):
92 mL
Standard Taper Joint:
45/12
Price (SGD)
2,067.00
PK
Add to cart
Tall, cylindrical form with standard taper caps
With Outside standard taper Ground Caps
Caps protect bottle in case of tipping; have open bottom
Prevent material from touching cap edges, provide increased headroom
Designed from Fed. Spec. NNN-B-1493, Type I, Style II
Specifications
Capacity
3.11 oz., 92mL
Capacity (Metric)
92 mL
Capacity (English)
3.11 oz.
Diameter (English) Inner
1.57 in.
Diameter (Metric) Inner
40 mm
Form
Tall Form and Regular
Material
Glass
Standard Taper Joint
45/12
Weight (Metric)
83g
Weight (English)
0.18 lb.
Shape
Cylindrical
Height (Metric)
100 mm
Height (English)
3.93 in.
Closure Material
Glass
Closure Type
Cap