Epoxy Coated Steel Wire Rack
Actual product may vary
Epoxy Coated Steel Wire Rack
Holds 15 conical bottom 15 mL centrifuge tubes.
Have Questions?
Catalog NumberPositions
FIS03-422-30615
Catalog number FIS03-422-306
Price (SGD)
106.00
Each
Positions:
15
Price (SGD)
106.00
Each
  • Top and middle sections have 18 mm square holes and are centered over rails in the base that hold the tips of the tubes
  • Generously spaced for easy handling
  • Heat-cured epoxy coating is unaffected by organic solvents, salts, weak acids and weak alkalis
  • Temperature range -223°C (-369°F) to 121°C (250°F); autoclavable
Specifications
AutoclavableAutoclavable
ColorBlue
HoldsTubes (15 mL), Tubes (16 to 20 mm)
MaterialEpoxy-coated Steel Wire
Positions15
ShapeRectangular
StackableYes, When Empty
SterilityNon-sterile
Diameter (Metric) Opening20 mm
Product TypeCentrifuge Tube Rack
Shelves3 Tiers
Array5 x 3
Unit SizeEach