Weighing Bottles, Low, Flat Form
Weighing Bottles, Low, Flat Form
Actual product may vary
Weighing Bottles, Low, Flat Form
Rugged glass bottles
Have Questions?
Catalog NumberCapacity (Metric)
FIS03-420A35 mL
Catalog number FIS03-420A
Price (SGD)
145.00
EA
Add to cart
Capacity (Metric):
35 mL
Price (SGD)
145.00
EA
Add to cart
Rugged glass bottles
  • Lamp-blown glass
  • Sturdy bottoms prevent chipping and facilitate cleaning
  • Tall, cylindrical form weighing bottles also available (Cat. Nos. 03-415B, -C, -E and -H)
Specifications
Capacity1.18 oz., 35mL
Capacity (Metric)35 mL
Capacity (English)1.18 oz.
ColorClear
Diameter (English) Inner1.96 in.
Diameter (Metric) Inner50 mm
FormLow Form
MaterialGlass
SterilityNon-sterile
Standard Taper Joint50/12
Weight (Metric)53g
Weight (English)0.11 lb.
ShapeRound
Height (Metric)30 mm
Product TypeWeighing Bottle
Height (English)1.18 in.
Closure TypePlug
Unit SizeEA