Weighing Bottles, Tall, Cylindrical Form
Weighing Bottles, Tall, Cylindrical Form
Actual product may vary
Weighing Bottles, Tall, Cylindrical Form
Rugged glass bottles
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric) Inner
FIS03-415H70 mL40 mm
Catalog number FIS03-415H
Price (SGD)
1,148.00
EA
Add to cart
Capacity (Metric):
70 mL
Diameter (Metric) Inner:
40 mm
Price (SGD)
1,148.00
EA
Add to cart
  • Lamp-blown glass
  • Sturdy bottoms prevent chipping and facilitate cleaning
  • Low, flat form weighing bottles also available (Cat. Nos. 03-420A through -420C)
Specifications
Capacity2.36 oz., 70mL
Capacity (Metric)70 mL
Capacity (English)2.36 oz.
ColorClear
Diameter (English) Inner1.57 in.
Diameter (Metric) Inner40 mm
FormTall Form
MaterialGlass
SterilityNon-sterile
Standard Taper Joint40/12
Weight (Metric)67g
Weight (English)0.14 lb.
ShapeCylindrical
Height (Metric)80 mm
Product TypeWeighing Bottle
Height (English)3.19 in.
Closure TypeCap
Unit SizeEA