Benchtop Biohazard Waste Disposal Bag
Benchtop Biohazard Waste Disposal Bag
Actual product may vary
Benchtop Biohazard Waste Disposal Bag
Benchtop Biohazard Waste Disposal Bag
Benchtop Biohazard Waste Disposal Bag
Fisherbrand
Benchtop Biohazard Waste Disposal Bag
Bright red, autoclavable bags feature one long flap for easy opening and disposal of tips and tubes
Have Questions?
Catalog NumberColor
FIS03-410-591Red
Catalog number FIS03-410-591
Price (SGD)
78.00
PK
Add to cart
Color:
Red
Price (SGD)
78.00
PK
Add to cart
Bright red, autoclavable bags feature one long flap for easy opening and disposal of tips and tubes
  • Size allows the bag to be inverted over user's hand to safely pick up waste
  • Printed with the biohazard symbol and sterilization indicator patch
  • Constructed from High Molecular Weight, High-Density (HMHD) polyethylene
  • Steam autoclavable at 121°C (250°F)
  • For space-saving waste disposal, use with Poxygrid™ Benchtop Biohazard Bag Holder (Mfr. No. H13193-1000)
Specifications
DesignPrinted Biohazard Symbols
Capacity (English)0.4 gal.
ColorRed
DisposableSingle-use
Height (Metric)28 cm
Width (Metric)22 cm
AutoclavableAutoclavable
For Use With (Application)Easy Opening and Disposal of Tips and Tubes
Width (English)8.5 in.
Thickness0.07 mm
Height (English)11 in.
MaterialHDPE
Product TypeBiohazard Waste Bag (Benchtop)
Unit SizePK