Narrow Neck Reagent Bottle
Actual product may vary
Narrow Neck Reagent Bottle
Universal bottles for storing, packing, taking samples, sampling and transport of liquids.
Have Questions?
Catalog NumberCapacity (Metric)Height (Metric)
FIS03-400-500500 mL170 mm
Catalog number FIS03-400-500
Price (SGD)
10.00
Each
Capacity (Metric):
500 mL
Height (Metric):
170 mm
Price (SGD)
10.00
Each
  • HDPE, transparent
  • PP tamper-evident screw cap, blue
Specifications
Capacity (Metric)500 mL
Height (Metric)170 mm
Length (Metric)74 mm
Unit SizeEach