11mm Wide Opening Snap-It Caps and Septa are easy to apply and easy to remove and fully compatible with all Snap-It vials.
Have Questions?
Catalog Number
Color
Septum
ColorSeal
FIS03-396Y
Blue, White
Blue PTFE/White Silicone, Pre-slit
Catalog number FIS03-396Y
Price (SGD)
-
Color:
Blue, White
Septum:
Blue PTFE/White Silicone, Pre-slit
Eliminate the need for crimping or de-capping tools with Thermo Scientific™ 11mm Wide Opening Snap-It Caps and Septa.They are easy to apply and easy to remove and fully compatible with all Snap-It vials. The enlarged open area allows for needle penetration across the entire vial opening. These chemically inert polyethylene caps are suitable for most chromatography applications and are an economical choice for routine HPLC, GC and IC applications, but with a low sealing property and zero resealing capacity. Choose from Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ brands.
Easy application and removal
Improved sealing against sample evaporation
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description11mm Snap-It Cap, 6mm hole
ColorBlue, White
MaterialPTFE, Silicone, Polyethylene
SeptumBlue PTFE/White Silicone, Pre-slit
Product LineNational Snap-It
Quantity100
Thickness (Metric)1.0 mm
TypeSnap Cap
Closure Size11 mm
For Use With (Equipment)National 11 mm Snap-It Vials