Manual Crimpers, Decrimpers and Decrimping Pliers
Actual product may vary
Thermo Scientific
Manual Crimpers, Decrimpers and Decrimping Pliers
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Have Questions?
Catalog NumberSizeType
FIS03-375-8A11 mmManual Decrimper
Catalog number FIS03-375-8A
Price (SGD)
-
Size:
11 mm
Type:
Manual Decrimper
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.
Manual Crimpers
Lightweight, precision crimpers attach seals to chromatography or serum vials. Available for standard aluminum or flip-off seals.
  • Factory set to properly crimp standard-thickness seals
  • Adjustable handle setting screw allows proper crimps with septa of varying thickness
Manual Decrimpers
Forged pliers feature a fitted gripping surface designed to allow removal of the listed sizes of seals.
  • Simply grip pliers around top of seal, squeeze and pry upward to remove
Manual Decrimping Pliers
Place decrimping head squarely on seal and squeeze.
  • Minimizes damage to vial rim with chromatography or serum vials
  • Removes standard aluminum seals and many steel seals
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionManual Decrimper
No. per Pack1 Ea.
TypeManual Decrimper
For Use With (Equipment)11 mm Standard Seal, National Vial, Chromacol Vial
Size11 mm
For Use With (Application)Industrial Chromatography
Product LineNational
Unit SizeEach