Kimble™ 20mL Glass Screw-Thread Scintillation Vials
Actual product may vary
DWK Life Sciences
Kimble™ 20mL Glass Screw-Thread Scintillation Vials
Manufactured from clear KG-33 borosilicate glass
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterial (Liner)Closure MaterialClosure TypeClosure Size
FIS03-340-021Foamed PolyethylenePolypropyleneAttached Screw Cap24 mm
FIS03-340-4CPulp-backed FoilPolypropyleneAttached Screw Cap22 mm
FIS03-340-4DFoamed PolyethylenePolypropyleneAttached Screw Cap22 mm
FIS03-340-4FPoly Seal ConeUreaScrew Cap22 mm
FIS03-340-4LPoly Seal ConeUreaAttached Screw Cap22 mm
FIS03-340-027Metal FoilPolypropyleneScrew Cap24 mm
FIS22-043-725Foil/CorkUreaAttached Screw Cap22 mm
Catalog number FIS03-340-021
Price (SGD)
-
Material (Liner):
Foamed Polyethylene
Closure Material:
Polypropylene
Closure Type:
Attached Screw Cap
Closure Size:
24 mm
Manufactured from clear KG-33 borosilicate glass
Variety of cap materials and liners available.
Specifications
Material (Liner)Foamed Polyethylene
ColorWhite
MaterialBorosilicate Glass
Volume (Metric)20 mL
For Use With (Application)Fast and easy identification
Packaging TypeCellular Trays
Volume (English)0.67 oz.
Closure MaterialPolypropylene
Closure TypeAttached Screw Cap
Closure Size24 mm
Certifications/ComplianceASTM E438
Unit SizeCase