Polystyrene Containers
Actual product may vary
Polystyrene Containers
Polystyrene Containers
Polystyrene Containers
Polystyrene Containers
Ideal for storing dry materials, powders and tablets.
Have Questions?
Catalog NumberDiameter (English) InnerVolume (English)Quantity
FIS03-338-3B0.86 in.5 dr.144/Cs.
Catalog number FIS03-338-3B
Price (SGD)
151.00
Case
Diameter (English) Inner:
0.86 in.
Volume (English):
5 dr.
Quantity:
144/Cs.
Price (SGD)
151.00
Case
Ideal for storing dry materials, powders and tablets.
  • Made of polystyrene material
  • With snap-on caps
Specifications
Diameter (English) Inner0.86 in.
Diameter (Metric) Inner22 mm
ColorClear
MaterialPolystyrene
Volume (Metric)18.5 mL
For Use With (Application)Storing dry materials, powders, tablets
StyleCylindrical
Temperature Range (English)158°F
Temperature Range (Metric)70°C
Volume (English)5 dr.
Volume (Metric) Total18.5 mL
Closure MaterialPolyethylene
Closure TypeSnap Cap
Quantity144/Cs.
Height (Metric)48 mm
Product TypeStorage Vial with Cap
Unit SizeCase