Nalgene™ Square Wide-Mouth Large PPCO Bottle with Closure: Autoclavable
Nalgene™ Square Wide-Mouth Large PPCO Bottle with Closure: Autoclavable
Actual product may vary
Nalgene™ Square Wide-Mouth Large PPCO Bottle with Closure: Autoclavable
Nalgene™ Square Wide-Mouth Large PPCO Bottle with Closure: Autoclavable
Nalgene™ Square Wide-Mouth Large PPCO Bottle with Closure: Autoclavable
Thermo Scientific
Nalgene™ Square Wide-Mouth Large PPCO Bottle with Closure: Autoclavable
Autoclave and store contaminated slides and other small lab equipment with the Thermo Scientific™ Nalgene™ Square Wide-Mouth Large PPCO Bottle with Closure.
Have Questions?
Catalog NumberCapacity (Metric)
FIS03-3134 L
Catalog number FIS03-313
Price (SGD)
133.00
EA
Add to cart
Capacity (Metric):
4 L
Price (SGD)
133.00
EA
Add to cart
Autoclave and store contaminated slides and other small lab equipment with the Thermo Scientific™ Nalgene™ Square Wide-Mouth Large PPCO Bottle with Closure. This polypropylene copolymer bottle is excellent for collecting, storing and transporting dry chemicals and environmental samples. Space saving design features molded-in handgrips for greater convenience and safety when pouring.
Features:
  • Polypropylene screw closures included
  • Wide mouth makes it excellent for collecting, storing and transporting dry chemicals and environmental samples
  • Bottle is not liquid leakproof†
  • Manufactured from non-toxic materials
Note:
Completely disengage threads or remove cap before autoclaving.
Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  These bottles and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene PP and PPCO bottles if services are available in your community.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity (Metric)4 L
Capacity (English)1 gal.
ColorTranslucent
DescriptionLarge Wide-Mouth Square Bottle
Diameter (English)5.6 in.
Diameter (English) Inner Neck3.5 in.
Diameter (Metric) Inner Neck89 mm
MaterialPPCO
No. per Case6
No. per Pack1
SterilityNon-sterile
Closure ColorWhite
Certifications/ComplianceUSP Class VI; FDA - 21 CFR
Closure IncludedYes
AutoclavableAutoclavable
GraduatedNo
MouthWide
ShapeSquare
Height (Metric)282 mm
Product TypeWide Mouth Bottle (Large)
Width (English)5.66 in.
Height (English)11.1 in.
Closure MaterialPolypropylene
Closure Size100 mm
Diameter (Metric)142 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizeEA