PYREX™ Gas Washing, Tall-Form, Fritted Cylinder Bottles
PYREX™ Gas Washing, Tall-Form, Fritted Cylinder Bottles
Actual product may vary
PYREX™ Gas Washing, Tall-Form, Fritted Cylinder Bottles
Tall design permits thorough washing.
Have Questions?
Catalog NumberDiameter (English) OutsideCapacity (English)Porosity
FIS03-040C2.48 in.16 oz.Coarse
Catalog number FIS03-040C
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (English) Outside:
2.48 in.
Capacity (English):
16 oz.
Porosity:
Coarse
Stopper is equipped with a 12 mm diameter fritted cylinder instead of a disc as the distributor.
Specifications
Capacity (Metric)473.17 mL
Capacity (English)16 oz.
MaterialBorosilicate Glass
TypeTall Form Gas Washing Bottle
Includes0.47 in. O.D. coarse porosity fritted disc and stopper assembly
PorosityCoarse
Standard Taper Joint29/42
SterilityNon-sterile
Diameter (English) Outside2.48 in.
Diameter (Metric) Outside6.3 cm
For Use With (Application)Provide an inexpensive but effective method for washing and drying gases
QuantityEach
Unit SizeEA