Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Diameter (Metric) Outside | Includes | Pore Size |
---|---|---|---|---|
FIS03-038F | 473.17 mL | 6.4 cm | 0.75 in. O.D. extra coarse porocity fritted disc and stopper assembly | 170 to 220 μm |
FIS03-038E | 236.58 mL | 5.1 cm | 0.75 in. O.D. extra coarse porocity fritted disc and stopper assembly | 170 to 220 μm |
These bottles have a large hexagonal base for stability. The fritted disc provides greater efficiency and more uniform dispersion of gas bubbles for complete absorption. Tubulation O.D. is approximately 8 mm.