Polymethylpentene Graduated Cylinders
Actual product may vary
Fisherbrand
Polymethylpentene Graduated Cylinders
Calibrated To Contain/To Deliver at 20°C
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FIS03-007-361000 mL10 mL
Catalog number FIS03-007-36
Price (SGD)
38.00
Each
Capacity (Metric):
1000 mL
Graduations:
10 mL
Price (SGD)
38.00
Each
Calibrated To Contain/To Deliver at 20°C
  • Transparent cylinders with large molded-in graduations
  • Easy to read
  • Large rounded bases ribbed for reinforcement and stability
  • Generous pour spout; 10mL size has a conical top for easier filling and pouring
  • Non-wetting
  • Chemical, heat and impact resistant
  • Window-clear
Specifications
Capacity (Metric)1000 mL
MaterialPMP
CalibrationTo Contain, To Deliver
Graduations10 mL
AutoclavableAutoclavable
Subdivision10mL
Base TypeCircular
Product TypeGraduated Cylinder
Unit SizeEach