Round and Oval Dropper Bottles with Attached Dropper Assemblies
Round and Oval Dropper Bottles with Attached Dropper Assemblies
Actual product may vary
Qorpak
Round and Oval Dropper Bottles with Attached Dropper Assemblies
Used to drip solvent
Have Questions?
Catalog NumberCapacity (Metric)ColorShapeQuantity
FIS02-992-545120 mLAmberRound24/Cs.
Catalog number FIS02-992-545
Price (SGD)
405.00
CS
Add to cart
Capacity (Metric):
120 mL
Color:
Amber
Shape:
Round
Quantity:
24/Cs.
Price (SGD)
405.00
CS
Add to cart
  • Space saving design
  • Available with caps made of different materials (Polypropylene hole and phenolic hole)
  • Available in amber and clear color
Specifications
Capacity4.05 oz., 120 mL
Capacity (Metric)120 mL
Capacity (English)4.05 oz.
ColorAmber
Diameter (English)1.88 in.
MaterialGlass
Quantity24/Cs.
Closure ColorBlack
ShapeRound
Height (Metric)112 mm
Height (English)4.4 in.
Closure MaterialPolypropylene
Closure Size20-400 mm
Diameter (Metric)48 mm
Closure TypeScrew Cap
Unit SizeCS