▸
Pipet Products
▸
Dropping Pipets
▸
Kimble™ Droppers
Actual product may vary
DWK Life Sciences
Kimble™ Droppers
With black rubber nipple
Have Questions?
Change view
Catalog Number
Volume (Metric)
FIS02-992-1B
60 mL
FIS02-992-1A
30 mL
2 Options
Catalog number
FIS02-992-1B
Price (SGD)
67.00
Case
Add to cart
Volume (Metric):
60 mL
Price (SGD)
67.00
Case
Add to cart
Bottles not included
Specifications
Material
Glass
Includes
Black Rubber nipple
Volume (Metric)
60 mL
Type
Dropper
For Use With (Equipment)
Dropping Bottle, 60 mL