Kimble™ KIMAX™ Brand Pipet Dropping Bottle
Kimble™ KIMAX™ Brand Pipet Dropping Bottle
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Brand Pipet Dropping Bottle
Medium length pipet dropper
Have Questions?
Catalog NumberCapacity (Metric)Standard Taper Joint
FIS02-990A30 mL12/18
Catalog number FIS02-990A
Price (SGD)
1,894.00
CS
Add to cart
Capacity (Metric):
30 mL
Standard Taper Joint:
12/18
Price (SGD)
1,894.00
CS
Add to cart
Medium length pipet dropper
  • Rubber nipple
  • KG-33 ASTM 438 Type 1 Class A Glass
  • Dropper Capacity: 1.0mL
Specifications
Capacity1 oz., 30mL
Capacity (Metric)30 mL
Capacity (English)1 oz.
ColorClear
Diameter (English)1.37 in.
MaterialBorosilicate Glass
Standard Taper Joint12/18
Dropper Capacity1 mL
ShapeRound
Height (Metric)113 mm
Product TypePipet Dropping Bottle
Width (English)1.37 in.
Height (English)4.44 in.
Diameter (Metric)35 mm
Through the Closure DispensingYes
Unit SizeCS