PYREX™ Dropping Bottles with Bulb and Pipet
PYREX™ Dropping Bottles with Bulb and Pipet
Actual product may vary
PYREX™ Dropping Bottles with Bulb and Pipet
Recommended for use when preventing contamination by solubility or pH change is important.
Have Questions?
Catalog NumberCapacity (Metric)
FIS02-986C125 mL
Catalog number FIS02-986C
Price (SGD)
2,598.00
CS
Add to cart
Capacity (Metric):
125 mL
Price (SGD)
2,598.00
CS
Add to cart
  • Constructed of Pyrex borosilicate glass
  • Interchangeable 1:10 taper
  • Closure type: hollow stopper
Specifications
Capacity (Metric)125 mL
Diameter (English)2.16 in.
IncludesBulb and Pipet
MaterialBorosilicate Glass
No. per Case12/Cs.
No. per Pack12/Pk.
Certifications/ComplianceASTM D-281
Height (Metric)150 mm
Height (English)5.9 in.
Closure MaterialGlass
Diameter (Metric)55 mm
Closure TypeHollow Stopper
Unit SizeCS