Check Order Status
Wheaton™ Redi-Cap™ Phenolic Screw Caps for Redi-Pak™ Containers
Actual product may vary
DWK Life Sciences
Wheaton™ Redi-Cap™ Phenolic Screw Caps for Redi-Pak™ Containers
Unique cone liner design offers superior torque retention and excellent sealing characteristics
Have Questions?
Catalog Number
Neck Finish
FIS02-984-23
24-400
Catalog number
FIS02-984-23
Price (SGD)
155.00
Case
Add to cart
Neck Finish:
24-400
Price (SGD)
155.00
Case
Add to cart
Poly-Seal liner is manufactured from low density polyethylene
Phenolic
Screw cap
Color: Black
Specifications
Color
Black
Material
Phenolic Resin
No. per Case
144 Cs.
Neck Finish
24-400
Certifications/Compliance
ISO 9001
For Use With (Equipment)
Redi-pak™ container
Closure Liner
Polyseal