Kimble™ KIMAX™ Aspirator Bottles with Tubulation
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Aspirator Bottles with Tubulation
Bottom glass hose-connection outlet is fused to bottle
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Stopper Number
FIS02-973-1E5000 mL10
FIS02-973-1B500 mL4
FIS02-973-1D2000 mL6
FIS02-973-1F10 L12
Catalog number FIS02-973-1E
Price (SGD)
-
Capacity (Metric):
5000 mL
Stopper Number:
10
Bottom glass hose-connection outlet is fused to bottle
  • With tooled top
  • Narrow mouth for rubber stopper
  • With white enamel marking spot
Specifications
Capacity (Metric)5000 mL
Diameter (English)7.2 in.
Diameter (English) Inner Neck1.65 in.
Diameter (Metric) Inner Neck42 mm
For Use With (Application)Designed to store and discharge liquids via a bottom hose outlet
IncludesBottom hose outlet, Marking Spot
MaterialBorosilicate Glass
Stopper Number10
MouthNarrow
Height (Metric)320 mm
Height (English)12.6 in.
Diameter (Metric)183 mm
Diameter (English) Inner Tubing0.312 in.
Diameter (Metric) Inner Tubing7.92 mm
Capacity (English)1.25 gal.
Product TypeReservoir Bottle
Unit SizeCase