Kimble™ KIMAX™ Aspirator Bottles with Tubulation
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Aspirator Bottles with Tubulation
Bottom glass hose-connection outlet is fused to bottle
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Stopper Number
FIS02-973-1B500 mL4
FIS02-973-1D2000 mL6
FIS02-973-1E5000 mL10
FIS02-973-1F10 L12
Catalog number FIS02-973-1B
Price (SGD)
345.00
Case
Capacity (Metric):
500 mL
Stopper Number:
4
Price (SGD)
345.00
Case
Bottom glass hose-connection outlet is fused to bottle
  • With tooled top
  • Narrow mouth for rubber stopper
  • With white enamel marking spot
Specifications
Capacity (Metric)500 mL
Capacity (English)0.125 gal.
Diameter (English)3.5 in.
Diameter (English) Inner Neck0.86 in.
Diameter (Metric) Inner Neck22 mm
For Use With (Application)Designed to store and discharge liquids via a bottom hose outlet
IncludesBottom hose outlet, Marking Spot
MaterialBorosilicate Glass
Stopper Number4
MouthNarrow
Height (Metric)165 mm
Product TypeReservoir Bottle
Height (English)6.5 in.
Diameter (Metric)89 mm
Diameter (English) Inner Tubing0.312 in.
Diameter (Metric) Inner Tubing7.92 mm
Unit SizeCase