Nalgene™ Polypropylene, Quick Action Spigot
Actual product may vary
Nalgene™ Polypropylene, Quick Action Spigot
Nalgene™ Polypropylene, Quick Action Spigot
Nalgene™ Polypropylene, Quick Action Spigot
Thermo Scientific
Nalgene™ Polypropylene, Quick Action Spigot
For Use With: Nalgene carboys
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
FIS02-963-7Spigot
6422-0010Spigot
Catalog number FIS02-963-7
Price (SGD)
155.00
Each
Product Type:
Spigot
Price (SGD)
155.00
Each
Stop drips and keep your Nalgene carboys in service for years with these Thermo Scientific™ Nalgene™ Polypropylene Quick-Action Replacement Spigots. Includes an HDPE tubing adapter which accepts tubing with 1/4 or 5/16 in. I.D for added versatility.
Features:
  • Spigot without tubing adapter accepts 5/8″ (1.6cm) I.D. tubing
  • Supplied with HDPE adapter for use with 1/4" or 5/16″ (0.6 or 0.8cm) tubing
Notes:
  • Disassemble spigot before autoclaving; tubing adapter is not autoclavable
Specifications
DescriptionQuick-Action Spigot PP fits 1-1/8 in. spigot boss for carboys 1-1/8 inch
Diameter (English) Inner Tubing5/8 in. (includes adapter for 1/4 or 5/16 in. tubing)
Diameter (Metric) Inner Tubing1.6 cm (includes adapter for 0.6 or 0.8 cm tubing)
For Use With (Equipment)Nalgene Carboys
MaterialPolypropylene
No. per Pack1 Ea.
Product TypeSpigot
Unit SizeEach