Nalgene™ LDPE, Round Carboy
Actual product may vary
Thermo Scientific
Nalgene™ LDPE, Round Carboy
For Use With: Store and transport reagents; Closure Material: Polypropylene; Color: White
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS02-961D10 L
FKS1079270125 L
Catalog number FIS02-961D
Price (SGD)
308.00
Each
Capacity (Metric):
10 L
Price (SGD)
308.00
Each
Store and transport reagents with the Thermo Scientific™ Nalgene™ Round LDPE Carboys, featuring wide shoulder handles that allow for easy carrying and pouring with gloved hands. These carboys accept 13.5 rubber stoppers for an alternative sealing method.
  • Molded-in graduations of 5 L and 1 gal. increments
  • Leakproof†
Includes:White polypropylene screw cap and 83B closure
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
AutoclavableNot Autoclavable
Capacity (English)2.6 gal.
Capacity (Metric)10 L
Closure MaterialPolypropylene
Closure Size83B
ColorWhite
Diameter (Metric) Inner Neck64 mm
Diameter (Metric) Outer250 mm
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
Height (Metric) With Closure389 mm
IncludesHandles
MaterialLDPE
No. per Case6/Cs.
No. per Pack1 Ea.
Product TypeCarboy with Handle
ShapeRound
SterilityNon-sterile
Workflow StepUpstream process solution storage. Upstream Validation
Unit SizeEach