Nalgene™ Polypropylene, Rectangular Carboy with Closure
Actual product may vary
Thermo Scientific
Nalgene™ Polypropylene, Rectangular Carboy with Closure
For Use With: Store solutions and handle large volumes of powders and other solid samples; Autoclavable; Closure Material: Polypropylene; Color: White
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS02-961-53A9 L
FIS02-961-53C20 L
Catalog number FIS02-961-53A
Price (SGD)
465.00
Each
Capacity (Metric):
9 L
Price (SGD)
465.00
Each
Store solutions and handle large volumes of powders and other solid samples with Thermo Scientific™ Nalgene™ Rectangular Polypropylene Carboys with Closure. Featuring a space-saving design, a wide mouth opening for filling and cleaning and a sturdy stainless-steel handle, these carboys are tough, translucent and autoclavable.
Features:
  • Wide-mouth opening for easy filling, transferring and cleaning of carboys
  • Molded in graduations in 1 gallon and 5 L increments for ease in dispensing
  • Stainless-steel handle for ease in transporting containers
  • Autoclavable, graduated and leakproof†
Includes:
  • Polypropylene screw closure
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
AutoclavableAutoclavable
Capacity (English)2.4 gal.
Capacity (Metric)9 L
Closure MaterialPolypropylene
Closure Size100-415 mm
ColorWhite
Dimensions (L x W x H)9 x 6 x 14 in. (220 x 153 x 360 mm)
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
MaterialPolypropylene
No. per Case6/Cs.
No. per Pack1 Ea.
Product TypeRectangular Carboy
SterilityNon-sterile
Workflow StepUpstream process solution storage. Upstream Validation
Unit SizeEach