Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure
Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure
Actual product may vary
Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure
Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure
Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure
Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure
Thermo Scientific
Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure
Store corrosive chemicals and distilled water with these large, narrow-mouth PPCO bottles, which are resistant to most acids, bases and alcohols.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FIS02-924C8 L53B
FIS02-924B4 L38-430 mm
2203-00208 L53B
Catalog number FIS02-924C
Price (SGD)
230.00
EA
Add to cart
Capacity (Metric):
8 L
Closure Size:
53B
Price (SGD)
230.00
EA
Add to cart
Store corrosive chemicals and distilled water with Thermo Scientific™ Nalgene™ Large Narrow-Mouth PPCO Bottles with Closure, which are resistant to most acids, bases and alcohols. Large sizes of these polypropylene copolymer bottles have a built-in shoulder loop.
Features:
  • 4 and 8L feature a built-in shoulder loop on larger sizes for attaching identification tag
  • Lighter weight than glass alternatives make for easy lifting
  • Translucence allows for easy viewing of liquid levels
  • Linerless caps ensure leakproof† protection without the use of liner that can wrinkle, cause leaks, or contaiminate your reagents
  • Autoclavable/Leakproof† when used with Nalgene closures
  • Manufactured from non-toxic materials
Includes:
Polypropylene screw caps
Note:
Completely disengage threads or remove closure before autoclaving. Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.
Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  These bottles and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene PP and PPCO bottles if services are available in your community.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity2 gal., 8 L
Capacity (Metric)8 L
Capacity (English)2 gal.
ColorTranslucent
DescriptionLarge Narrow-Mouth Bottle
Diameter (Metric) Inner Neck37 mm
MaterialPPCO
No. per Case6
No. per Pack1
SterilityNon-sterile
Closure ColorWhite
Neck Finish53B
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure IncludedYes
AutoclavableAutoclavable
DisposableReusable
GraduatedNo
MouthNarrow
ShapeRound
Height (Metric)409 mm
Product TypeNarrow Mouth Bottle
Width (English)7.6 in.
Height (English)16.1 in.
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size53B
Diameter (Metric)193 mm
Closure TypePolypropylene Screw Closure w/TPE Gasket
Through the Closure DispensingNo
Unit SizeEA