Nalgene™ Rectangular HDPE Bottles with Closure
Nalgene™ Rectangular HDPE Bottles with Closure
Actual product may vary
Nalgene™ Rectangular HDPE Bottles with Closure
Nalgene™ Rectangular HDPE Bottles with Closure
Nalgene™ Rectangular HDPE Bottles with Closure
Nalgene™ Rectangular HDPE Bottles with Closure
Thermo Scientific
Nalgene™ Rectangular HDPE Bottles with Closure
Transport, sample or store liquids and dry materials with these space-efficient bottles featuring excellent chemical resistance to most acids, bases and alcohols.
Have Questions?
Catalog NumberCapacity (Metric)Closure Size
FIS02-924-5C250 mL38 mm
Catalog number FIS02-924-5C
Price (SGD)
141.00
PK
Add to cart
Capacity (Metric):
250 mL
Closure Size:
38 mm
Price (SGD)
141.00
PK
Add to cart
Transport, sample or store liquids and dry materials with these space-efficient bottles that feature excellent chemical resistance to most acids, bases and alcohols. Thermo Scientific™ Nalgene™ Rectangular HDPE Bottles with Closure take up less shelf space than traditional round bottles. HDPE bottles have a sturdy wall construction with straight sides and a wide mouth for easy filling. Made only from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
  • Excellent chemical resistance to most acids, bases and alcohols make these excellent containers to protect your valuable samples
  • Rectangular shape makes it easy to fit in freezer for storage up to -100°C (-148°F)
  • Linerless polypropylene screw closures provide leakproof† protection without the use of a liner that can wrinkle, cause leaks, or contaminate your reagents
  • Leakproof† when used with Nalgene closures
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
Includes:
Polypropylene screw caps
Recycling Information: Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.
.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity8 oz., 250 mL
Capacity (Metric)250 mL
Capacity (English)8 oz.
ColorTranslucent
DescriptionWide-Mouth Rectangular Bottle
Diameter (English) Inner Neck1.1 in.
Diameter (Metric) Inner Neck28 mm
MaterialHDPE
Length (Metric)76 mm
No. per Case72
No. per Pack12
SterilityNon-sterile
Width (Metric)51 mm
Closure ColorNatural, translucent
Certifications/ComplianceUSP Class VI; FDA - 21 CFR
Closure IncludedYes
GraduatedNo
MouthWide
ShapeRectangular
ACT LabelYes
Height (Metric)117 mm
Product TypeWide Mouth Bottle
Height (English)4.6 in.
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size38 mm
Diameter (Metric)76.2 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizePK