Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures
Actual product may vary
Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures
Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures
Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures
Thermo Scientific
Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures
Fit these replacement closures onto Thermo Scientific™ Nalgene™ narrow-mouth and wide-mouth bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberThread Size (Metric)Product TypeColor
FIS02-923-14DD43 mmReplacement CapNatural
712160-038438 mmReplacement CapNatural
712150-053053 mmReplacement CapNatural
Catalog number FIS02-923-14DD
Price (SGD)
97.00
Pack
Thread Size (Metric):
43 mm
Product Type:
Replacement Cap
Color:
Natural
Price (SGD)
97.00
Pack
Fit Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures onto Nalgene narrow-mouth and wide-mouth bottles.
  • Made to fit Nalgene narrow-mouth and wide-mouth bottles
  • Leakproof† when used with the Nalgene bottles with the matching finish
.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
ColorNatural
DescriptionBottle Replacement Closures
MaterialPolypropylene
No. per Case12
Thread Size (Metric)43 mm
MouthWide
ShapeRound
Product TypeReplacement Cap
For Use With (Equipment)Nalgene narrow-mouth and wide-mouth bottles
Unit SizePack