Wheaton™ Safety-Coated Amber Bottles with PTFE-faced Rubber Lined Cap
Wheaton™ Safety-Coated Amber Bottles with PTFE-faced Rubber Lined Cap
Actual product may vary
Wheaton™ Safety-Coated Amber Bottles with PTFE-faced Rubber Lined Cap
Wheaton™ Safety-Coated Amber Bottles with PTFE-faced Rubber Lined Cap
DWK Life Sciences
Wheaton™ Safety-Coated Amber Bottles with PTFE-faced Rubber Lined Cap
Create a safe work environment with Wheaton Safety Coated Bottles. If a bottle is accidentally broken, the PVC based plastisol coating on the outside of bottle retains the contents long enough for proper disposal.
Have Questions?
Catalog NumberCapacity (Metric)
FIS02-918D1000 mL
Catalog number FIS02-918D
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
  • Ideal for lab or field applications, the bottle's non-slip coating allows for greater ease and stability when handling wet or dry
  • Bottles are manufactured from soda lime glass that conforms to USP Type III requirements
  • 8, 16 and 32oz Boston Round style
  • 80oz and 4L with jug handle
Specifications
Capacity (Metric)1000 mL
Capacity (English)32 oz.
ColorAmber
Diameter (English)3.7 in.
MaterialGlass
Closure ColorBlack
Height (Metric)210 mm
Product TypeSafety-coated Bottle
Height (English)8.2 in.
Closure LinerPTFE/Rubber
Closure MaterialPhenolic
Closure Size33-400 mm
Diameter (Metric)96 mm
Closure TypeScrew Cap
Unit SizeCS