Certified Cleaned Straight Sided Bottles
Actual product may vary
Certified Cleaned Straight Sided Bottles
Certified Cleaned Straight Sided Bottles
Certified Cleaned Straight Sided Bottles
Certified Cleaned Straight Sided Bottles
Ideal for soil sampling and environmental applications. Fisherbrand™ Certified Cleaned Straight Sided Bottles are manufactured from clear soda glass.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS02-912-312500 mL
FIS02-912-310125 mL
FIS02-912-3131 L
Catalog number FIS02-912-312
Price (SGD)
346.00
Case
Capacity (Metric):
500 mL
Price (SGD)
346.00
Case
Ideal for soil sampling and environmental applications. Fisherbrand™ Certified Cleaned Straight Sided Bottles are manufactured from clear soda glass.
  • Cleaned per EPA guidelines for semi-volatiles, pesticides, PCBs and metals
  • Straight sided walls allow for the complete removal of contents
  • Bottles are autoclavable; caps are not
Specifications
Capacity (Metric)500 mL
Capacity (English)16 oz.
ColorClear
MaterialSoda Lime Glass
Length (Metric)55mm
Quantity12/PK.
Closure ColorWhite
Liner MaterialPTFE
Certifications/ComplianceCertificate of analysis
Closure IncludedYes
Closure Size89-400 mm
Closure TypeScrew Cap
Unit SizeCase