▸
Bottles, Jars and Jugs
▸
Bottles
▸
Environmental Sampling Bottles and Accessories
▸
Environmental Sampling Bottles
▸
Custom Cleaned Amber Wide Mouth Packer Bottles
Actual product may vary
Custom Cleaned Amber Wide Mouth Packer Bottles
Ideal storage for light sensitive dry powders. Fisherbrand™ Custom Cleaned Amber Wide Mouth Packer Bottles are manufactured using amber soda glass.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Length (Metric)
Quantity
FIS02-912-003
500 mL
121mm
12/PK.
FIS02-912-004
30 mL
24/PK.
FIS02-911-930
60 mL
101mm
24/PK.
FIS02-911-933
250 mL
101mm
24/PK.
4 Options
Catalog number
FIS02-912-003
Price (SGD)
238.00
Case
Add to cart
Capacity (Metric):
500 mL
Length (Metric):
121mm
Quantity:
12/PK.
Price (SGD)
238.00
Case
Add to cart
Ideal storage for light sensitive dry powders. Fisherbrand™ Custom Cleaned Amber Wide Mouth Packer Bottles are manufactured using amber soda glass.
Wide mouth allows for easy accessibility
Pre-cleaned for visible particulates using deionised water
With white polypropylene cap with PTFE faced foamed polyethylene liner
Specifications
Capacity (Metric)
500 mL
Capacity (English)
16.9 oz.
Cleaning Procedure
EPA guidelines
Color
Amber
Material
Glass
Length (Metric)
121mm
Quantity
12/PK.
Closure Color
White
Liner Material
PTFE
Certifications/Compliance
EPA
Closure Included
Yes
Mouth
Wide
Product Type
Wide Mouth Bottle
Closure Liner
Polyethylene
Closure Material
PTFE
Closure Size
53-400 mm
Closure Type
Screw Cap