Easy-Squeeze Wash Bottles, 6/Cs.
Actual product may vary
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy-Squeeze Wash Bottles, 6/Cs.
Easy to fill and ready to use
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorChemical Label
FIS02-897-3BlueDeionized Water
FIS02-897-11BlueWater
FIS02-897-4BlueDistilled Water
FIS02-897-10NaturalUniversal
FIS02-897-5NaturalEthanol
FIS02-897-1RedAcetone
Catalog number FIS02-897-3
Price (SGD)
124.00
Case
Color:
Blue
Chemical Label:
Deionized Water
Price (SGD)
124.00
Case
Easy to fill and ready to use
  • Wide mouth
  • Translucent and sputter-free
  • With easy-to-read color-coded silk-screened labels
  • Tips can be cut back to increase flow
  • When not in use, bottles relieve internal pressure without dripping
Label information includes:
  • Chemical name
  • National Fire Protection Association (NFPA) codes and symbols
  • Chemical Abstracts Service (CAS) number
  • Hazard codes
  • Laboratory protective equipment information
  • Target organ information
  • Recommended fill level
  • LDPE recycling code
  • Bottle volume capacity “For Dispensing Only, Not Recommended For Storage” statement
Not recommended for use above 60°C (140°F).
Specifications
Capacity17 oz., 500mL
Capacity (Metric)500 mL
ColorBlue
Diameter (English)2.99 in.
IncludesPTFE membrane
MaterialLDPE
Closure ColorBlue
Closure IncludedYes
GraduatedNo
MouthWide
Height (Metric)173 mm
Height (English)6.81 in.
Chemical LabelDeionized Water
Closure MaterialPolypropylene
Closure Size38-415 mm
Diameter (Metric)76 mm
Closure TypePolypropylene Closure
Through the Closure DispensingNo
AutoclavableNot Autoclavable
Capacity (English)17 oz.
ShapeCylindrical
Certifications/ComplianceISO 13485:2003
Product TypeWash Bottle
Unit SizeCase